Assay Details
Target Gene Details
Entrez Gene ID: | 64344 |
Gene Name: | hypoxia inducible factor 3 alpha subunit |
Gene Aliases: |
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17 |
Location: |
Chr.19:46297046-46343433 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
More Information
Set Membership: |
|

Genomic Map