Assay Details
Target Gene Details
Entrez Gene ID: | 50944 |
Gene Name: | SH3 and multiple ankyrin repeat domains 1 |
Gene Aliases: |
SPANK-1, SSTRIP, synamon |
Location: |
Chr.19:50659257-50719779 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHANK1 | NM_016148.3 | 23 | 7158 | NP_057232.2 |
| XM_006723233.3 | 24 | 7556 | XP_006723296.1 | |
| XM_011527013.2 | 25 | 7580 | XP_011525315.1 | |
| XM_011527014.2 | 23 | 7529 | XP_011525316.1 | |
| AY461451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3574587 | Chr.19:50613167 - 50689920 on Build GRCh38 | Gain |
|
| nsv828583 | Chr.19:50637495 - 50746580 on Build GRCh38 | Loss |
|
| nsv579950 | Chr.19:50645515 - 50690186 on Build GRCh38 | Loss |
|
| nsv953605 | Chr.19:50659244 - 50662943 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map