Assay Details
Target Gene Details
Entrez Gene ID: | 26664 |
Gene Name: | olfactory receptor family 7 subfamily C member 1 |
Gene Aliases: |
CIT-HSP-146E8, HSTPCR86P, OR19-5, OR7C4, TPCR86 |
Location: |
Chr.19:14797550-14835376 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR7C1 | XM_005259853.3 | XP_005259910.1 | ||
| XM_011527915.2 | XP_011526217.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115378091] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map