Assay Details
Target Gene Details
Entrez Gene ID: | 59285 |
Gene Name: | calcium voltage-gated channel auxiliary subunit gamma 6 |
Gene Aliases: |
- |
Location: |
Chr.19:53991149-54012666 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNG6 | NM_031897.2 | NP_114103.2 | ||
| NM_145814.1 | NP_665813.1 | |||
| NM_145815.1 | NP_665814.1 | |||
| NR_102308.1 | ||||
| XM_006723316.3 | XP_006723379.1 | |||
| AF288386.1 | AAK20029.1 | |||
| AF361352.1 | AAL50047.1 | |||
| BC021552.1 | AAH21552.1 | |||
| BX281606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv580110 | Chr.19:53961144 - 54093354 on Build GRCh38 | Gain |
|
| nsv1057516 | Chr.19:53986233 - 54046901 on Build GRCh38 | Gain |
|
| nsv1063127 | Chr.19:53926876 - 54068977 on Build GRCh38 | Gain |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
| nsv833877 | Chr.19:53951529 - 54117323 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map