Assay Details
Target Gene Details
Entrez Gene ID: | 83639 |
Gene Name: | testis expressed 101 |
Gene Aliases: |
CT131, GTPR867, NYD-SP8, PRO1884, SGRG, SPATA44, TES101RP |
Location: |
Chr.19:43388611-43418615 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEX101 | NM_001130011.1 | NP_001123483.1 | ||
| NM_031451.4 | NP_113639.4 | |||
| XM_005259303.3 | XP_005259360.2 | |||
| AF241268.1 | AAK28327.1 | |||
| AF353396.1 | AAK38662.1 | |||
| AY014285.1 | AAK27310.1 | |||
| AY359077.1 | AAQ89436.1 | |||
| BC001861.2 | AAH01861.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469666 | Chr.19:43272995 - 43430182 on Build GRCh38 | Loss |
|
| nsv1057949 | Chr.19:43380906 - 43431018 on Build GRCh38 | Loss |
|
| esv3583431 | Chr.19:43412826 - 43449282 on Build GRCh38 | Loss |
|
| esv3644466 | Chr.19:43383591 - 43418114 on Build GRCh38 | Gain |
|
| dgv3603n100 | Chr.19:43304322 - 43590925 on Build GRCh38 | Gain |
|
| nsv1060280 | Chr.19:43362671 - 43455533 on Build GRCh38 | Gain |
|
| nsv1057657 | Chr.19:42798345 - 43572412 on Build GRCh38 | Gain |
|
| nsv509744 | Chr.19:43400292 - 43476817 on Build GRCh38 | Insertion |
|
| dgv3604n100 | Chr.19:43414688 - 43453235 on Build GRCh38 | Loss |
|
| nsv520852 | Chr.19:43380933 - 43455962 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73563824] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map