Assay Details
Target Gene Details
Entrez Gene ID: | 23373 |
Gene Name: | CREB regulated transcription coactivator 1 |
Gene Aliases: |
MECT1, TORC-1, TORC1, WAMTP1 |
Location: |
Chr.19:18683615-18782333 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRTC1 | NM_001098482.1 | NP_001091952.1 | ||
| NM_015321.2 | NP_056136.2 | |||
| XM_005259833.3 | XP_005259890.1 | |||
| XM_005259834.1 | XP_005259891.1 | |||
| XM_005259835.3 | XP_005259892.1 | |||
| XM_005259836.3 | XP_005259893.1 | |||
| XM_006722710.3 | XP_006722773.1 | |||
| XM_011527842.2 | XP_011526144.1 | |||
| XM_017026536.1 | XP_016882025.1 | |||
| XM_017026537.1 | XP_016882026.1 | |||
| XM_017026538.1 | XP_016882027.1 | |||
| AB014516.1 | BAA31591.1 | |||
| AK024089.1 | BAB14822.1 | |||
| AU118537.1 | ||||
| AY040323.1 | AAK93832.1 | |||
| AY360171.1 | AAQ98856.1 | |||
| BC028050.1 | AAH28050.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
| nsv952422 | Chr.19:18606691 - 18807891 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78988515] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map