Assay Details
Target Gene Details
Entrez Gene ID: | 8570 |
Gene Name: | KH-type splicing regulatory protein |
Gene Aliases: |
FBP2, FUBP2, KSRP |
Location: |
Chr.19:6413104-6424811 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KHSRP | NM_003685.2 | NP_003676.2 | ||
| XM_005259668.4 | 19 | 2488 | XP_005259725.1 | |
| XM_011528395.2 | 20 | 2453 | XP_011526697.1 | |
| XM_017027408.1 | 20 | 2377 | XP_016882897.1 | |
| XM_017027409.1 | XP_016882898.1 | |||
| XM_017027410.1 | 19 | 2347 | XP_016882899.1 | |
| AK300957.1 | ||||
| BC085004.1 | AAH85004.1 | |||
| U69126.1 | AAC50892.1 | |||
| U94832.1 | AAB53222.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833727 | Chr.19:6400101 - 6568241 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map