Assay Details
Target Gene Details
Entrez Gene ID: | 8193 |
Gene Name: | double PHD fingers 1 |
Gene Aliases: |
BAF45b, NEUD4, neuro-d4 |
Location: |
Chr.19:38211006-38229714 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPF1 | NM_001135155.2 | 12 | 2140 | NP_001128627.1 |
| NM_001135156.2 | 12 | 2154 | NP_001128628.1 | |
| NM_001289978.1 | 12 | 2170 | NP_001276907.1 | |
| NM_004647.3 | 11 | 2038 | NP_004638.2 | |
| XM_005259289.2 | 11 | 2089 | XP_005259346.1 | |
| XM_005259290.3 | 12 | 2111 | XP_005259347.1 | |
| XM_005259292.4 | 11 | 2026 | XP_005259349.1 | |
| XM_006723407.2 | 12 | 2141 | XP_006723470.1 | |
| XM_006723408.3 | 12 | 2109 | XP_006723471.1 | |
| XM_006723409.2 | 12 | 2156 | XP_006723472.1 | |
| XM_006723410.2 | 9 | 1708 | XP_006723473.1 | |
| XM_011527356.1 | 11 | 2119 | XP_011525658.1 | |
| XM_011527357.2 | 11 | 2059 | XP_011525659.1 | |
| XM_011527358.2 | 12 | 2185 | XP_011525660.1 | |
| AI857563.1 | 1 | 190 | ||
| AI885717.1 | 1 | 209 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833821 | Chr.19:38139297 - 38266152 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map