Assay Details
Target Gene Details
Entrez Gene ID: | 79156 |
Gene Name: | pleckstrin homology and FYVE domain containing 1 |
Gene Aliases: |
APPD, LAPF, PHAFIN1, ZFYVE15 |
Location: |
Chr.19:29665420-29675477 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHF1 | NM_024310.4 | NP_077286.3 | ||
| XM_005259256.3 | XP_005259313.1 | |||
| XM_011527309.2 | XP_011525611.1 | |||
| AK027758.1 | BAB55349.1 | |||
| AK225814.1 | ||||
| AK301616.1 | ||||
| AY037145.2 | AAK67626.2 | |||
| BC002744.2 | AAH02744.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061604 | Chr.19:28373028 - 29773397 on Build GRCh38 | Loss |
|
| nsv833799 | Chr.19:29550565 - 29683695 on Build GRCh38 | Gain |
|
| nsv1061174 | Chr.19:29668772 - 29760417 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map