Assay Details
Target Gene Details
Entrez Gene ID: | 84705 |
Gene Name: | GTP binding protein 3 (mitochondrial) |
Gene Aliases: |
COXPD23, GTPBG3, MSS1, MTGP1, THDF1 |
Location: |
Chr.19:17334982-17342731 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP3 | NM_001128855.2 | 9 | 2443 | NP_001122327.1 |
| NM_001195422.1 | 9 | 2499 | NP_001182351.1 | |
| NM_032620.3 | 9 | 2506 | NP_116009.2 | |
| NM_133644.3 | 8 | 2602 | NP_598399.2 | |
| AK027606.1 | 9 | 2453 | BAB55228.1 | |
| AK091035.1 | 9 | 2824 | ||
| AY078988.1 | 9 | 2468 | AAL85493.1 | |
| BC008924.2 | 7 | 1983 | ||
| BC019261.1 | 9 | 2447 | AAH19261.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833768 | Chr.19:17338840 - 17526738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map