Assay Details
Target Gene Details
Entrez Gene ID: | 7138 |
Gene Name: | troponin T1, slow skeletal type |
Gene Aliases: |
ANM, NEM5, STNT, TNT, TNTS |
Location: |
Chr.19:55132698-55149354 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNNT1 | NM_001126132.2 | NP_001119604.1 | ||
| NM_001126133.2 | NP_001119605.1 | |||
| NM_001291774.1 | NP_001278703.1 | |||
| NM_003283.5 | NP_003274.3 | |||
| XM_011527246.2 | XP_011525548.1 | |||
| XM_017027186.1 | XP_016882675.1 | |||
| XM_017027187.1 | XP_016882676.1 | |||
| AK291352.1 | ||||
| AK308199.1 | ||||
| AY762903.1 | AAX07831.1 | |||
| AY762904.1 | AAX07832.1 | |||
| BC007074.1 | ||||
| BC010963.1 | AAH10963.1 | |||
| BC022086.2 | AAH22086.2 | |||
| BC034143.1 | AAH34143.1 | |||
| BC107798.1 | AAI07799.1 | |||
| BT019630.1 | AAV38436.1 | |||
| DC385627.1 | ||||
| DC385695.1 | ||||
| M19308.1 | AAA61205.1 | |||
| M19309.1 | AAA61204.1 | |||
| S51938.1 | ||||
| S69208.1 | AAB30272.1 | |||
| S69209.1 | AAB30273.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs146534494] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map