Assay Details
Target Gene Details
Entrez Gene ID: | 9230 |
Gene Name: | RAB11B, member RAS oncogene family |
Gene Aliases: |
H-YPT3 |
Location: |
Chr.19:8390321-8404434 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB11B | NM_004218.3 | NP_004209.2 | ||
| AF498947.1 | AAM21095.1 | |||
| AK225036.1 | ||||
| AK225642.1 | ||||
| AK297498.1 | ||||
| AK310573.1 | ||||
| AK312994.1 | ||||
| BC110081.1 | AAI10082.1 | |||
| BP367041.1 | ||||
| BT019535.1 | AAV38342.1 | |||
| BT019536.1 | AAV38343.1 | |||
| CR536494.1 | CAG38733.1 | |||
| CR541691.1 | CAG46492.1 | |||
| D38516.1 | ||||
| EF560724.1 | ||||
| X79780.1 | CAA56176.1 |
More Information
Set Membership: |
|

Genomic Map