Assay Details
Target Gene Details
Entrez Gene ID: | 199692 |
Gene Name: | zinc finger protein 627 |
Gene Aliases: |
- |
Location: |
Chr.19:11575255-11619159 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF627 | XM_011527780.2 | XP_011526082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509721 | Chr.19:11525216 - 11691390 on Build GRCh38 | Insertion |
|
| esv991647 | Chr.19:11575212 - 11587674 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map