Assay Details
Target Gene Details
Entrez Gene ID: | 80271 |
Gene Name: | inositol-trisphosphate 3-kinase C |
Gene Aliases: |
IP3-3KC, IP3KC |
Location: |
Chr.19:40717103-40740860 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITPKC | NM_025194.2 | NP_079470.1 | ||
| XM_017027324.1 | XP_016882813.1 | |||
| AJ290975.1 | CAC40815.1 | |||
| AK024596.1 | ||||
| BC060788.1 | AAH60788.1 | |||
| D38169.1 | BAA22524.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160619 | Chr.19:40706352 - 40763204 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114760337,rs141182844] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map