Assay Details
Target Gene Details
Entrez Gene ID: | 4670 |
Gene Name: | heterogeneous nuclear ribonucleoprotein M |
Gene Aliases: |
CEAR, HNRNPM4, HNRPM, HNRPM4, HTGR1, NAGR1, hnRNP M |
Location: |
Chr.19:8444575-8489118 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
More Information
Set Membership: |
|

Genomic Map