Assay Details
Target Gene Details
Entrez Gene ID: | 147991 |
Gene Name: | dpy-19 like 3 (C. elegans) |
Gene Aliases: |
- |
Location: |
Chr.19:32405749-32485893 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPY19L3 | NM_001172774.1 | NP_001166245.1 | ||
| NM_207325.2 | NP_997208.2 | |||
| XM_006723045.1 | XP_006723108.1 | |||
| XM_011526526.2 | XP_011524828.1 | |||
| XM_017026366.1 | XP_016881855.1 | |||
| XM_017026367.1 | XP_016881856.1 | |||
| AK126264.1 | BAC86508.1 | |||
| AK126757.1 | BAC86676.1 | |||
| AK129497.1 | BAC85166.1 | |||
| BC131725.1 | ||||
| CR749459.1 | CAH18293.1 |
More Information
Set Membership: |
|

Genomic Map