Assay Details
Target Gene Details
Entrez Gene ID: | 84705 |
Gene Name: | GTP binding protein 3 (mitochondrial) |
Gene Aliases: |
COXPD23, GTPBG3, MSS1, MTGP1, THDF1 |
Location: |
Chr.19:17334982-17342731 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP3 | NM_001195422.1 | NP_001182351.1 | ||
| DC339966.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3643818 | Chr.19:17331902 - 17338827 on Build GRCh38 | Gain |
|
| nsv470129 | Chr.19:17188084 - 17341718 on Build GRCh38 | Loss |
|
| esv21604 | Chr.19:17332440 - 17339470 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map