Assay Details
Target Gene Details
Entrez Gene ID: | 83639 |
Gene Name: | testis expressed 101 |
Gene Aliases: |
CT131, GTPR867, NYD-SP8, PRO1884, SGRG, SPATA44, TES101RP |
Location: |
Chr.19:43388611-43418615 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEX101 | NM_031451.4 | NP_113639.4 | ||
| AF241268.1 | AAK28327.1 | |||
| BC001861.2 | AAH01861.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469666 | Chr.19:43272995 - 43430182 on Build GRCh38 | Loss |
|
| nsv1057949 | Chr.19:43380906 - 43431018 on Build GRCh38 | Loss |
|
| esv3644466 | Chr.19:43383591 - 43418114 on Build GRCh38 | Gain |
|
| dgv3603n100 | Chr.19:43304322 - 43590925 on Build GRCh38 | Gain |
|
| nsv1057657 | Chr.19:42798345 - 43572412 on Build GRCh38 | Gain |
|
| nsv1060280 | Chr.19:43362671 - 43455533 on Build GRCh38 | Gain |
|
| nsv509744 | Chr.19:43400292 - 43476817 on Build GRCh38 | Insertion |
|
| nsv520852 | Chr.19:43380933 - 43455962 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map