Assay Details
Target Gene Details
Entrez Gene ID: | 5348 |
Gene Name: | FXYD domain containing ion transport regulator 1 |
Gene Aliases: |
PLM |
Location: |
Chr.19:35137207-35143055 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FXYD1 | XM_017026874.1 | XP_016882363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953282 | Chr.19:35134797 - 35153797 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map