Assay Details
Target Gene Details
Entrez Gene ID: | 9230 |
Gene Name: | RAB11B, member RAS oncogene family |
Gene Aliases: |
H-YPT3 |
Location: |
Chr.19:8390321-8404434 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB11B | NM_004218.3 | 5 | 1175 | NP_004209.2 |
| BP396219.1 | ||||
| CX787570.1 | 3 | 726 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578558 | Chr.19:8403267 - 8411965 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map