Assay Details
Target Gene Details
Entrez Gene ID: | 4670 |
Gene Name: | heterogeneous nuclear ribonucleoprotein M |
Gene Aliases: |
CEAR, HNRNPM4, HNRPM, HNRPM4, HTGR1, NAGR1, hnRNP M |
Location: |
Chr.19:8444575-8489118 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
More Information
Additional Information:
For this assay, SNP(s) [rs141784669,rs147598953] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map