Assay Details
Target Gene Details
Entrez Gene ID: | 64857 |
Gene Name: | pleckstrin homology and RhoGEF domain containing G2 |
Gene Aliases: |
ARHGEF42, CLG, LDAMD |
Location: |
Chr.19:39412582-39428415 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHG2 | XM_006723334.2 | 1 | 361 | XP_006723397.1 |
| XM_017027150.1 | 1 | 361 | XP_016882639.1 | |
| XM_017027151.1 | 1 | 361 | XP_016882640.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114784999] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map