Assay Details
Target Gene Details
Entrez Gene ID: | 54815 |
Gene Name: | GATA zinc finger domain containing 2A |
Gene Aliases: |
p66alpha |
Location: |
Chr.19:19385833-19508932 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GATAD2A | NM_017660.3 | NP_060130.3 | ||
XM_011528105.1 | XP_011526407.1 | |||
XM_017026903.1 | XP_016882392.1 | |||
XM_017026905.1 | XP_016882394.1 | |||
XM_017026906.1 | XP_016882395.1 | |||
XM_017026907.1 | XP_016882396.1 | |||
XM_017026908.1 | XP_016882397.1 | |||
XM_017026909.1 | XP_016882398.1 | |||
XM_017026911.1 | XP_016882400.1 | |||
AK303482.1 | ||||
BG773264.1 | ||||
DB123918.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
