Assay Details
Target Gene Details
Entrez Gene ID: | 126410 |
Gene Name: | cytochrome P450 family 4 subfamily F member 22 |
Gene Aliases: |
ARCI5, INLNE, LI3 |
Location: |
Chr.19:15508487-15552317 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4F22 | NM_173483.3 | NP_775754.2 | ||
| XM_011527692.2 | XP_011525994.1 | |||
| XM_011527693.2 | XP_011525995.1 | |||
| AK096820.1 | BAC04868.1 | |||
| DA874258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057788 | Chr.19:15497917 - 15594886 on Build GRCh38 | Loss |
|
| nsv1064684 | Chr.19:15311319 - 16062011 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map