Assay Details
Target Gene Details
Entrez Gene ID: | 84902 |
Gene Name: | centrosomal protein 89 |
Gene Aliases: |
CCDC123, CEP123 |
Location: |
Chr.19:32875925-32972029 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP89 | NM_032816.4 | NP_116205.3 | ||
| XM_005259344.3 | XP_005259401.1 | |||
| XM_011527425.2 | XP_011525727.1 | |||
| AK027546.1 | BAB55190.1 | |||
| AL832158.1 | ||||
| BC136328.1 | ||||
| BU628988.1 | ||||
| DB539713.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062926 | Chr.19:32828718 - 32904288 on Build GRCh38 | Loss |
|
| esv24841 | Chr.19:32879638 - 32880273 on Build GRCh38 | Loss |
|
| nsv516922 | Chr.19:32844072 - 32883216 on Build GRCh38 | Loss |
|
| nsv1060014 | Chr.19:32854920 - 33011720 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75674520] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map