Assay Details
Target Gene Details
Entrez Gene ID: | 24139 |
Gene Name: | echinoderm microtubule associated protein like 2 |
Gene Aliases: |
ELP70, EMAP-2, EMAP2 |
Location: |
Chr.19:45609396-45645601 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EML2 | NM_001193268.1 | 2 | 328 | NP_001180197.1 |
| NM_001193269.1 | 2 | 232 | NP_001180198.1 | |
| XM_006723125.2 | 2 | 412 | XP_006723188.1 | |
| XM_006723126.2 | 2 | 220 | XP_006723189.1 | |
| XM_011526694.2 | 2 | 412 | XP_011524996.1 | |
| XM_011526695.2 | 2 | 412 | XP_011524997.1 | |
| XM_017026551.1 | 2 | 412 | XP_016882040.1 | |
| AK294668.1 | 2 | 279 | ||
| AK295905.1 | 2 | 232 | ||
| AK296258.1 | 2 | 161 | ||
| AK302961.1 | 2 | 292 | ||
| AK308580.1 | 2 | 160 |
Target Gene Details
Entrez Gene ID: | 100287177 |
Gene Name: | EML2 antisense RNA 1 |
Gene Aliases: |
C19orf83 |
Location: |
Chr.19:45641494-45642840 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EML2-AS1 | NR_130728.1 | 3 | 513 | |
| NR_130729.1 | 2 | 319 | ||
| BI334767.1 | 2 | 319 | ||
| CA424540.1 | 1 | 581 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833848 | Chr.19:45457372 - 45644939 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map