Assay Details
Target Gene Details
Entrez Gene ID: | 390937 |
Gene Name: | Ets2 repressor factor-like |
Gene Aliases: |
- |
Location: |
Chr.19:41907549-41928721 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC390937 | XM_017027573.1 | XP_016883062.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv953580 | Chr.19:41888230 - 41997948 on Build GRCh38 | Deletion |
|
| dgv3550n100 | Chr.19:41916194 - 42001892 on Build GRCh38 | Gain |
|
| nsv1118619 | Chr.19:41915248 - 41921748 on Build GRCh38 | Duplication |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79600216] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map