Assay Details
Target Gene Details
Entrez Gene ID: | 7515 |
Gene Name: | X-ray repair cross complementing 1 |
Gene Aliases: |
RCC |
Location: |
Chr.19:43543312-43575578 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XRCC1 | NM_006297.2 | NP_006288.2 | ||
| AB208781.1 | BAD92018.1 | |||
| AK293542.1 | ||||
| AK300163.1 | ||||
| AK315332.1 | ||||
| BC023593.2 | AAH23593.1 | |||
| BP349667.1 | ||||
| CR456728.1 | CAG33009.1 | |||
| M36089.1 | AAA63270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3603n100 | Chr.19:43304322 - 43590925 on Build GRCh38 | Gain |
|
| nsv1056785 | Chr.19:43506218 - 43577449 on Build GRCh38 | Gain |
|
| nsv1057657 | Chr.19:42798345 - 43572412 on Build GRCh38 | Gain |
|
| nsv1063149 | Chr.19:43554929 - 43583431 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map