Assay Details
Target Gene Details
Entrez Gene ID: | 11283 |
Gene Name: | cytochrome P450 family 4 subfamily F member 8 |
Gene Aliases: |
CPF8, CYPIVF8 |
Location: |
Chr.19:15615218-15629638 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4F8 | NM_007253.3 | NP_009184.1 | ||
| XM_017026232.1 | XP_016881721.1 | |||
| AF133298.1 | AAD49566.1 | |||
| AK096809.1 | 2 | 1268 | ||
| AK300478.1 | 3 | 928 | ||
| AK300530.1 | ||||
| AK300539.1 | ||||
| AK314685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2718235 | Chr.19:15616082 - 15677993 on Build GRCh38 | Deletion |
|
| dgv3444n100 | Chr.19:15609874 - 15638182 on Build GRCh38 | Gain |
|
| dgv627e199 | Chr.19:15615949 - 15676977 on Build GRCh38 | Deletion |
|
| nsv1066963 | Chr.19:15601558 - 15642613 on Build GRCh38 | Gain |
|
| dgv3445n100 | Chr.19:15613287 - 15648121 on Build GRCh38 | Gain |
|
| dgv3443n100 | Chr.19:15542560 - 15644632 on Build GRCh38 | Gain |
|
| nsv1064684 | Chr.19:15311319 - 16062011 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map