Assay Details
Target Gene Details
Entrez Gene ID: | 2528 |
Gene Name: | fucosyltransferase 6 |
Gene Aliases: |
FCT3A, FT1A, Fuc-TVI, FucT-VI |
Location: |
Chr.19:5830626-5839753 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893160 | Chr.19:5827086 - 5839735 on Build GRCh38 | Loss |
|
| esv3643541 | Chr.19:5831170 - 5844611 on Build GRCh38 | Loss |
|
| nsv1062851 | Chr.19:5531540 - 5893590 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map