Assay Details
Target Gene Details
Entrez Gene ID: | 90522 |
Gene Name: | Yip1 interacting factor homolog B, membrane trafficking protein |
Gene Aliases: |
FinGER8 |
Location: |
Chr.19:38303560-38315976 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIF1B | NM_001039671.2 | NP_001034760.1 | ||
| NM_001039672.2 | NP_001034761.1 | |||
| NM_001039673.2 | NP_001034762.1 | |||
| NM_001145461.1 | NP_001138933.1 | |||
| NM_001145462.1 | NP_001138934.1 | |||
| NM_001145463.1 | NP_001138935.1 | |||
| XM_005259385.3 | XP_005259442.1 | |||
| XM_017027449.1 | XP_016882938.1 | |||
| XM_017027450.1 | XP_016882939.1 | |||
| XM_017027451.1 | XP_016882940.1 | |||
| AK304479.1 | ||||
| AL833382.1 | CAI46138.1 | |||
| AW630507.1 | ||||
| BC007644.1 | AAH07644.1 | |||
| BC014974.1 | AAH14974.1 | |||
| BC091477.1 | AAH91477.2 | |||
| BE877226.1 | ||||
| BG765536.1 | ||||
| BM703847.1 | ||||
| BX379501.2 | ||||
| CA487891.1 | ||||
| CA488303.1 | ||||
| CF127536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833822 | Chr.19:38270108 - 38454465 on Build GRCh38 | Loss |
|

Genomic Map