Assay Details
Target Gene Details
Entrez Gene ID: | 90075 |
Gene Name: | zinc finger protein 30 |
Gene Aliases: |
KOX28 |
Location: |
Chr.19:34923795-34945172 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF30 | XM_011527444.1 | XP_011525746.1 | ||
| XM_017027424.1 | XP_016882913.1 |
Target Gene Details
Entrez Gene ID: | 102723513 |
Gene Name: | ZNF30 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:34923015-34926826 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF30-AS1 | NR_110749.1 | |||
| HY123988.1 | ||||
| HY139988.1 |
More Information
Set Membership: |
|

Genomic Map