Assay Details
Target Gene Details
Entrez Gene ID: | 162989 |
Gene Name: | death effector domain containing 2 |
Gene Aliases: |
FLAME-3 |
Location: |
Chr.19:42198593-42220152 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEDD2 | NM_001270614.1 | NP_001257543.1 | ||
| NM_001270615.1 | NP_001257544.1 | |||
| NM_133328.3 | NP_579874.1 | |||
| NR_073046.1 | ||||
| NR_073047.1 | ||||
| NR_073048.1 | ||||
| NR_073049.1 | ||||
| NR_073050.1 | ||||
| NR_073051.1 | ||||
| NR_073052.1 | ||||
| XM_011526569.1 | XP_011524871.1 | |||
| XM_011526571.1 | XP_011524873.1 | |||
| XM_011526572.1 | XP_011524874.1 | |||
| XM_017026402.1 | XP_016881891.1 | |||
| XM_017026403.1 | XP_016881892.1 | |||
| AF443591.1 | AAL48220.1 | |||
| AF457575.1 | AAM10835.1 | |||
| AK075328.1 | BAC11551.1 | |||
| AK130203.1 | ||||
| AK307895.1 | ||||
| AY125488.1 | AAM95240.1 | |||
| BC013372.2 | AAH13372.2 | |||
| BC027930.1 | AAH27930.1 | |||
| BM908148.1 | ||||
| BX374729.2 | ||||
| BX458265.2 | ||||
| CN373297.1 | ||||
| CR984238.1 | ||||
| CT000394.1 | ||||
| HY033141.1 | ||||
| HY033487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss |
|
| nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain |
|
| nsv833833 | Chr.19:42153124 - 42278970 on Build GRCh38 | Loss |
|
| nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map