Assay Details
Target Gene Details
Entrez Gene ID: | 28954 |
Gene Name: | RRAD and GEM like GTPase 1 |
Gene Aliases: |
GD:REM, GES |
Location: |
Chr.20:31475273-31484905 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REM1 | NM_014012.5 | NP_054731.2 | ||
| XM_005260404.1 | XP_005260461.1 | |||
| XM_011528795.1 | XP_011527097.1 | |||
| XM_017027833.1 | XP_016883322.1 | |||
| AF084465.1 | AAC33132.1 | |||
| AF152863.1 | AAF74212.1 | |||
| AK313060.1 | ||||
| BC039813.1 | AAH39813.1 | |||
| BT020078.1 | AAV38881.1 | |||
| BT020079.1 | AAV38882.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470546 | Chr.20:31245806 - 31647499 on Build GRCh38 | Gain |
|
| esv2763228 | Chr.20:31247246 - 31651719 on Build GRCh38 | Gain |
|
| dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
|
| nsv458961 | Chr.20:31305698 - 31688103 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map