Assay Details
Target Gene Details
Entrez Gene ID: | 5740 |
Gene Name: | prostaglandin I2 (prostacyclin) synthase |
Gene Aliases: |
CYP8, CYP8A1, PGIS, PTGI |
Location: |
Chr.20:49503874-49568170 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGIS | NM_000961.3 | NP_000952.1 | ||
| AF297048.1 | AAG31781.1 | |||
| AF297049.1 | AAG31782.1 | |||
| AF297050.1 | AAG31783.1 | |||
| AF297051.1 | AAG31784.1 | |||
| AF297052.1 | AAG31785.1 | |||
| BC101809.1 | AAI01810.1 | |||
| BC101811.1 | AAI01812.1 | |||
| D38145.1 | BAA07343.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065852 | Chr.20:49502654 - 49643666 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map