Assay Details
Target Gene Details
Entrez Gene ID: | 23627 |
Gene Name: | prion protein 2 (dublet) |
Gene Aliases: |
DOPPEL, DPL, PrPLP, dJ1068H6.4 |
Location: |
Chr.20:4721854-4728462 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRND | NM_012409.3 | NP_036541.2 | ||
| AF187843.1 | AAG43448.1 | |||
| AK313404.1 | ||||
| AY358985.1 | AAQ89344.1 | |||
| BC043644.2 | AAH43644.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058830 | Chr.20:4663154 - 4725219 on Build GRCh38 | Gain |
|
| esv3645074 | Chr.20:4205961 - 5037636 on Build GRCh38 | Gain |
|
| nsv585310 | Chr.20:4586645 - 4794039 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map