Assay Details
Target Gene Details
Entrez Gene ID: | 6640 |
Gene Name: | syntrophin alpha 1 |
Gene Aliases: |
LQT12, SNT1, TACIP1, dJ1187J4.5 |
Location: |
Chr.20:33407957-33443892 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNTA1 | NM_003098.2 | NP_003089.1 | ||
| XM_005260517.1 | XP_005260574.1 | |||
| XM_011529007.1 | XP_011527309.1 | |||
| XM_011529008.1 | XP_011527310.1 | |||
| AK095942.1 | ||||
| AK291994.1 | ||||
| AK301800.1 | ||||
| BC026215.2 | AAH26215.1 | |||
| S81737.1 | AAB36398.1 | |||
| U40571.1 | AAC50448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064413 | Chr.20:33348381 - 33599978 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map