Assay Details
Target Gene Details
Entrez Gene ID: | 3745 |
Gene Name: | potassium voltage-gated channel subfamily B member 1 |
Gene Aliases: |
DRK1, Kv2.1 |
Location: |
Chr.20:49363877-49484033 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNB1 | NM_004975.3 | 2 | 10280 | NP_004966.1 |
| XM_006723784.3 | 3 | 10374 | XP_006723847.1 | |
| XM_011528799.2 | 3 | 10403 | XP_011527101.1 | |
| AK126976.1 | 1 | 2316 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064443 | Chr.20:49135574 - 49439352 on Build GRCh38 | Gain |
|
| nsv1067146 | Chr.20:48878894 - 49475527 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map