Assay Details
Target Gene Details
Entrez Gene ID: | 80025 |
Gene Name: | pantothenate kinase 2 |
Gene Aliases: |
C20orf48, HARP, HSS, NBIA1, PKAN |
Location: |
Chr.20:3888839-3929882 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PANK2 | NM_001324191.1 | NP_001311120.1 | ||
| NM_001324192.1 | NP_001311121.1 | |||
| NM_001324193.1 | NP_001311122.1 | |||
| NM_024960.5 | NP_079236.3 | |||
| NM_153638.3 | NP_705902.2 | |||
| NM_153640.3 | NP_705904.1 | |||
| NR_136715.1 | ||||
| XM_005260836.4 | XP_005260893.3 | |||
| XM_011529364.2 | XP_011527666.1 | |||
| XM_011529365.2 | XP_011527667.1 | |||
| XM_017028077.1 | XP_016883566.1 | |||
| XM_017028078.1 | 1 | 164 | XP_016883567.1 | |
| XM_017028079.1 | XP_016883568.1 | |||
| AF494409.1 | AAN32907.1 | |||
| AK021791.1 | BAB13897.1 | |||
| AK097796.1 | BAC05173.1 | |||
| AK310190.1 | ||||
| AL519408.3 | ||||
| AL532307.3 | ||||
| BC065019.1 | AAH65019.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645053 | Chr.20:3849918 - 3987827 on Build GRCh38 | Loss |
|
| nsv828676 | Chr.20:3898474 - 3902974 on Build GRCh38 | Loss |
|
| esv3645059 | Chr.20:3898170 - 3904490 on Build GRCh38 | Loss |
|
| dgv776e199 | Chr.20:3898151 - 3904682 on Build GRCh38 | Deletion |
|
| esv2722123 | Chr.20:3898160 - 3904528 on Build GRCh38 | Deletion |
|
| nsv828673 | Chr.20:3738382 - 4184811 on Build GRCh38 | Gain |
|
| esv3645060 | Chr.20:3899237 - 3901193 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map