Assay Details
Target Gene Details
Entrez Gene ID: | 146 |
Gene Name: | adrenoceptor alpha 1D |
Gene Aliases: |
ADRA1, ADRA1A, ADRA1R, ALPHA1, DAR, dJ779E11.2 |
Location: |
Chr.20:4220631-4249012 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADRA1D | NM_000678.3 | NP_000669.1 | ||
| L31772.1 | AAB59487.1 | |||
| S70782.1 | AAB31163.2 | |||
| U03864.1 | AAB60351.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645074 | Chr.20:4205961 - 5037636 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map