Assay Details
Target Gene Details
Entrez Gene ID: | 727993 |
Gene Name: | uncharacterized LOC727993 |
Gene Aliases: |
- |
Location: |
Chr.20:1947210-2007517 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC727993 | NR_134520.1 | |||
| AK090681.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|

Genomic Map