Assay Details
Target Gene Details
Entrez Gene ID: | 10005 |
Gene Name: | acyl-CoA thioesterase 8 |
Gene Aliases: |
HNAACTE, NAP1, PTE-1, PTE-2, PTE1, PTE2, hACTE-III, hTE |
Location: |
Chr.20:45841721-45857409 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOT8 | NM_005469.3 | NP_005460.2 | ||
| XM_005260239.3 | XP_005260296.1 | |||
| XM_011528480.2 | XP_011526782.1 | |||
| AF014404.1 | AAB71665.1 | |||
| AF124264.1 | AAD27616.1 | |||
| AK056668.1 | ||||
| AK126206.1 | ||||
| AK292494.1 | ||||
| AK296973.1 | ||||
| AK298783.1 | ||||
| BC117155.1 | ||||
| BC117157.1 | ||||
| BI911038.1 | ||||
| CR541771.1 | CAG46570.1 | |||
| CR541778.1 | CAG46577.1 | |||
| DB470803.3 | ||||
| X86032.1 | CAA60024.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458993 | Chr.20:45842587 - 45880235 on Build GRCh38 | Loss |
|
| nsv833990 | Chr.20:45771510 - 45946844 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map