Assay Details
Target Gene Details
Entrez Gene ID: | 6616 |
Gene Name: | synaptosome associated protein 25 |
Gene Aliases: |
CMS18, RIC-4, RIC4, SEC9, SNAP, SNAP-25, SUP, bA416N4.2, dJ1068F16.2 |
Location: |
Chr.20:10218694-10307420 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522874 | Chr.20:9918791 - 10838562 on Build GRCh38 | Loss |
|
| nsv3290 | Chr.20:10224314 - 10259651 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map