Assay Details
Target Gene Details
Entrez Gene ID: | 84969 |
Gene Name: | TOX high mobility group box family member 2 |
Gene Aliases: |
C20orf100, GCX-1, GCX1, dJ1108D11.2, dJ495O3.1 |
Location: |
Chr.20:43914852-44069616 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOX2 | NM_001098796.1 | NP_001092266.1 | ||
| NM_001098797.1 | NP_001092267.1 | |||
| NM_032883.2 | NP_116272.1 | |||
| AK289906.1 | ||||
| BM545579.1 | ||||
| CB989964.1 | ||||
| DA174963.1 | ||||
| JN655166.1 | ||||
| JN655167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057847 | Chr.20:42652464 - 44431352 on Build GRCh38 | Loss |
|
| nsv833989 | Chr.20:43746793 - 43947619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map