Assay Details
Target Gene Details
Entrez Gene ID: | 51497 |
Gene Name: | negative elongation factor complex member C/D |
Gene Aliases: |
HSPC130, NELF-C, NELF-D, TH1, TH1L |
Location: |
Chr.20:58981208-58995133 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NELFCD | NM_198976.2 | NP_945327.2 | ||
| AF161479.1 | AAF29094.1 | |||
| AJ238379.1 | CAB64339.1 | |||
| AK001316.1 | BAA91618.1 | |||
| AK023310.1 | BAB14519.1 | |||
| AK023927.1 | BAB14729.1 | |||
| AK293410.1 | ||||
| AK304310.1 | ||||
| BC014952.1 | AAH14952.1 | |||
| BI459250.1 | ||||
| BX647417.1 | ||||
| HY015074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525118 | Chr.20:58972890 - 58996708 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79087230] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map