Assay Details
Target Gene Details
Entrez Gene ID: | 140836 |
Gene Name: | barrier to autointegration factor 2 |
Gene Aliases: |
BAF-L, BAF2, BAFL, C20orf179 |
Location: |
Chr.20:17693675-17735872 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BANF2 | NM_001014977.3 | NP_001014977.2 | ||
| NM_001159495.1 | NP_001152967.1 | |||
| NM_178477.4 | NP_848572.3 | |||
| XM_005260668.3 | XP_005260725.1 | |||
| XM_011529170.1 | XP_011527472.1 | |||
| XM_011529171.2 | XP_011527473.1 | |||
| AA394107.1 | ||||
| AW022487.1 | ||||
| BC054871.1 | AAH54871.1 | |||
| BC062735.1 | AAH62735.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs73264783] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map