Assay Details
Target Gene Details
Entrez Gene ID: | 51378 |
Gene Name: | angiopoietin 4 |
Gene Aliases: |
ANG3, ANG4 |
Location: |
Chr.20:869899-916367 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPT4 | NM_001322809.1 | 8 | 3791 | NP_001309738.1 |
| NM_015985.3 | 9 | 3922 | NP_057069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510778 | Chr.20:843492 - 905972 on Build GRCh38 | Deletion |
|
| nsv1062082 | Chr.20:763183 - 928875 on Build GRCh38 | Gain |
|
| esv3644964 | Chr.20:846296 - 877968 on Build GRCh38 | Gain |
|
| esv3644952 | Chr.20:386013 - 1119183 on Build GRCh38 | Gain |
|
| nsv585191 | Chr.20:680149 - 913482 on Build GRCh38 | Gain |
|
| nsv1067424 | Chr.20:671381 - 911795 on Build GRCh38 | Gain |
|
| nsv585193 | Chr.20:842861 - 942563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map