Assay Details
Target Gene Details
Entrez Gene ID: | 164395 |
Gene Name: | tubulin tyrosine ligase like 9 |
Gene Aliases: |
C20orf125 |
Location: |
Chr.20:31870702-31944959 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3347 | Chr.20:31871679 - 31901524 on Build GRCh38 | Insertion |
|
| nsv833952 | Chr.20:31855501 - 32049500 on Build GRCh38 | Gain |
|
| dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map