Assay Details
Target Gene Details
Entrez Gene ID: | 343637 |
Gene Name: | R-spondin 4 |
Gene Aliases: |
C20orf182, CRISTIN4 |
Location: |
Chr.20:958452-1002264 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RSPO4 | NM_001029871.3 | NP_001025042.2 | ||
| NM_001040007.2 | NP_001035096.1 | |||
| XM_017027839.1 | XP_016883328.1 | |||
| AK122609.1 | ||||
| DQ355152.1 | ABC75877.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055853 | Chr.20:921359 - 1034331 on Build GRCh38 | Gain |
|
| esv3644952 | Chr.20:386013 - 1119183 on Build GRCh38 | Gain |
|
| esv274959 | Chr.20:961022 - 966160 on Build GRCh38 | Loss |
|
| nsv1067403 | Chr.20:925142 - 977237 on Build GRCh38 | Gain |
|
| esv3644966 | Chr.20:887288 - 978010 on Build GRCh38 | Gain |
|
| esv3575346 | Chr.20:884122 - 1168539 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73577400,rs77133653] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map