Assay Details
Target Gene Details
Entrez Gene ID: | 11317 |
Gene Name: | recombination signal binding protein for immunoglobulin kappa J region like |
Gene Aliases: |
RBPL, RBPSUHL, SUHL |
Location: |
Chr.20:45306836-45319677 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBPJL | NM_001281448.1 | NP_001268377.1 | ||
| NM_001281449.1 | NP_001268378.1 | |||
| NM_014276.3 | NP_055091.2 | |||
| XM_011528521.2 | XP_011526823.1 | |||
| XM_011528522.2 | XP_011526824.1 | |||
| XM_011528523.2 | XP_011526825.1 | |||
| AB024964.1 | BAA88232.1 | |||
| AB026048.1 | BAA86121.1 | |||
| AB027710.1 | BAA87051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645928 | Chr.20:45310422 - 45318758 on Build GRCh38 | Gain |
|
| nsv1056815 | Chr.20:45257396 - 45314077 on Build GRCh38 | Loss |
|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map